This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01494238_m1
See other IL31RA GE Assays ›
Gene Symbol:
Il31ra
Gene Name:
interleukin 31 receptor A
Gene Aliases:
-
Chromosome Location:
Chr.2: 44334661 - 44404217 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01494238_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il31ra
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257278.2 NP_001244207.1 12 - 13 1588 77
XM_008760740.1 XP_008758962.1 12 - 13 1763 77
GenBank mRNA JQ013736.1 - 12 - 13 1588 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002897.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s)

JAK-STAT cascade
positive regulation of cell proliferation
monocyte differentiation
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of tyrosine phosphorylation of Stat5 protein

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009260 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009260 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009260 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P276673 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200020385.rn.1 TC0200002897.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200020383.rn.1 TC0200002897.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200020382.rn.1 TC0200002897.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22294230 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1363530 - -
RatRef-12 Expression BeadChip ILMN_67924 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0