This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01494703_m1
See other NDUFC1 GE Assays ›
Gene Symbol:
Ndufc1
Gene Name:
NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 140460749 - 140464723 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01494703_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufc1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008761014.1 XP_008759236.1 2 - 3 342 154

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase

MicroArray Probes:

Agilent Affymetrix

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119093 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008077 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P008077 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P119093 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P008077 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P119093 - -
Array Name Probe IDs Targets Target Type
Rat Genome 230 2.0 Array 1392530_at - -
Rat Genome 230 2.0 Array 1392530_PM_at - -
Affymetrix HT Rat Focus Array 1392530_at - -
Affymetrix HT Rat Focus Array 1392530_PM_at - -
Rat Expression Set 230 1392530_PM_at - -
Rat Expression Set 230 1392530_at - -
Rat Genome U34 Set rc_AI104679_s_at - -
Rat Genome U34 Set rc_AI176491_at - -
Rat Genome U34 Set rc_AI176491_at - -
Rat Genome U34 Set rc_AI104679_s_at - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0