This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01495315_g1
See other GLMP GE Assays ›
Gene Symbol:
Glmp
Gene Name:
glycosylated lysosomal membrane protein
Gene Aliases:
0610031j06rik, RGD1303130
Chromosome Location:
Chr.2: 187697407 - 187701123 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01495315_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glmp
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001004226.1 NP_001004226.1 1 - 2 148 68
XM_008761133.1 XP_008759355.1 1 - 2 269 68
GenBank mRNA BC079294.1 - 1 - 2 148 7134483 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200001543.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Glmp

Gene Ontology Categories:

Function(s) Process(es)

intracellular receptor signaling pathway
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
ligand-dependent nuclear receptor transcription coactivator activity
transcription regulatory region DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200009696.rn.1 TC0200001543.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200009698.rn.1 TC0200001543.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0