Assay Details
| Gene Symbol: Smarcd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031983.2 | NP_114189.1 | 1 - 2 | 297 | 85 | |
| GenBank mRNA | BC062063.1 | - | 1 - 2 | 297 | 5599204 | 85 |
More Information
Updated Mapping Information
Set Membership: |
|

Smarcd2
Genomic Map