This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01496564_m1
See other NTHL1 GE Assays ›
Gene Symbol:
Nthl1
Gene Name:
nth (endonuclease III)-like 1 (E.coli)
Gene Aliases:
Nth1
Chromosome Location:
Chr.10: 13996660 - 14002827 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01496564_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nthl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001105728.1 NP_001099198.1 2 - 3 414 119

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000000214.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA catabolic process, endonucleolytic
base-excision repair, AP site formation
nucleotide-excision repair, DNA incision, 5'-to lesion
double-stranded DNA binding
DNA-(apurinic or apyrimidinic site) lyase activity
DNA N-glycosylase activity
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000002032.rn.1 TC1000000214.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000002036.rn.1 TC1000000214.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000002033.rn.1 TC1000000214.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000002035.rn.1 TC1000000214.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000002030.rn.1 TC1000000214.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline