This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01497125_m1
See other SPNS2 GE Assays ›
Gene Symbol:
Spns2
Gene Name:
spinster homolog 2
Gene Aliases:
-
Chromosome Location:
Chr.10: 59010428 - 59049482 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01497125_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spns2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144991.1 NP_001138463.1 1 - 2 548 88
XM_006246562.2 XP_006246624.2 1 - 2 545 88
XM_006246563.2 XP_006246625.2 1 - 2 545 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1000003180.rn.1

Set Membership:

Amplicon length between 86 and 100 Transporter Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

B cell homeostasis
lymphocyte homeostasis
regulation of humoral immune response
sphingosine-1-phosphate signaling pathway
sphingolipid metabolic process
lipid transport
locomotion
T cell homeostasis
regulation of eye pigmentation
lymph node development
transmembrane transport
bone development
lymphocyte migration
sphingolipid transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1000028699.rn.1 TC1000003180.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000028700.rn.1 TC1000003180.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000028698.rn.1 TC1000003180.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000028696.rn.1 TC1000003180.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000028701.rn.1 TC1000003180.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1000028697.rn.1 TC1000003180.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-9rjdb:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline