This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01499385_g1
See other UNG GE Assays ›
Gene Symbol:
Ung
Gene Name:
uracil-DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.12: 48246593 - 48256157 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01499385_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ung
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013124.1 NP_001013142.1 1 - 2 359 94
XM_008769336.1 XP_008767558.1 2 - 3 1030 94
GenBank mRNA BC091211.1 - 1 - 2 359 7381643 94

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ung

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
somatic hypermutation of immunoglobulin genes
somatic recombination of immunoglobulin gene segments
negative regulation of apoptotic process
uracil DNA N-glycosylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1200015368.rn.1 TC1200001786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200015369.rn.1 TC1200001786.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR1200015370.rn.1 TC1200001786.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0