This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01499869_m1
See other SDS GE Assays ›
Gene Symbol:
Sds
Gene Name:
serine dehydratase
Gene Aliases:
RATSDHE1, SDH2, Sdh, Sdhe1, TDH
Chromosome Location:
Chr.12: 41620429 - 41627741 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01499869_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sds
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053962.3 NP_446414.3 2 - 3 129 90
XM_008769213.1 XP_008767435.1 1 - 2 627 90
GenBank mRNA BC088110.1 - 2 - 3 129 7323391 90
Y00752.1 - 2 - 3 146 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC088110.1, NM_053962.3, XM_008769213.1, Y00752.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sds

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

gluconeogenesis
L-serine catabolic process
threonine catabolic process
response to nutrient levels
response to cobalamin
pyruvate biosynthetic process
response to amino acid
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
pyridoxal phosphate binding
protein homodimerization activity
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0