This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01500374_g1
See other PRSS37 GE Assays ›
Gene Symbol:
Prss37
Gene Name:
protease, serine, 37
Gene Aliases:
RGD1311823
Chromosome Location:
Chr.4: 68337498 - 68349286 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01500374_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prss37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108625.1 NP_001102095.1 1 - 2 112 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0400002746.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001108625.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
binding of sperm to zona pellucida
cell migration
protein maturation
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0400019605.rn.1 TC0400002746.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0400019604.rn.1 TC0400002746.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0