This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01502430_g1
See other POLR2K GE Assays ›
Gene Symbol:
Polr2k
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide K
Gene Aliases:
-
Chromosome Location:
Chr.7: 74939166 - 74992594 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01502430_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2k
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282325.1 NP_001269254.1 2 - 3 160 82
XM_006241527.2 XP_006241589.1 1 - 2 80 82
GenBank mRNA AW144706.1 - 2 - 3 160 82
EX488432.1 - 2 - 3 131 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
biological_process
molecular_function
DNA binding
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700007358.rn.1 TC0700000967.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700007354.rn.1 TC0700000967.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700007355.rn.1 TC0700000967.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700007357.rn.1 TC0700000967.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21967015 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0