This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01503180_m1
See other TMEM263 GE Assays ›
Gene Symbol:
Tmem263
Gene Name:
transmembrane protein 263
Gene Aliases:
RGD1563325
Chromosome Location:
Chr.7: 24647083 - 24668381 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01503180_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem263
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108739.1 NP_001102209.1 3 - 4 786 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700002343.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094458 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 844787 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 68838 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 418987 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 127338 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 758099 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 860342 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 627473 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278156 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 636074 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203683 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 797967 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 181295 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 347608 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 14326 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 148851 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094458 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094458 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094458 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P396422 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P155971 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 48423 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 3018 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10901291 - -
Affymetrix Rat Gene 1.0 ST Array 10901290 - -
Affymetrix Rat Gene 1.1 ST Array 10901289 - -
Rat Genome 230 2.0 Array 1383529_at - -
Rat Genome 230 2.0 Array 1383529_PM_at - -
Rat Genome 230 2.0 Array 1388789_at - -
Rat Genome 230 2.0 Array 1392942_at - -
Rat Genome 230 2.0 Array 1392942_PM_at - -
Rat Genome 230 2.0 Array 1388789_PM_at - -
Affymetrix HT Rat Focus Array 1392942_PM_at - -
Affymetrix HT Rat Focus Array 1392942_at - -
Affymetrix Rat Clariom S Assay PSR0700018713.rn.1 TC0700002343.rn.1 RTA TC
Rat Expression Set 230 1388789_PM_at - -
Rat Expression Set 230 1388789_at - -
Rat Expression Set 230 1383529_at - -
Rat Expression Set 230 1392942_PM_at - -
Rat Expression Set 230 1383529_PM_at - -
Rat Expression Set 230 1392942_at - -
Affymetrix Rat Gene 1.0 ST Array 10901289 - -
Rat Genome U34 Set rc_AI010326_at - -
Rat Genome U34 Set AA942842_at - -
Rat Genome U34 Set rc_AI103305_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22019903 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1362636 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0