This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01503181_m1
See other TMEM263 GE Assays ›
Gene Symbol:
Tmem263
Gene Name:
transmembrane protein 263
Gene Aliases:
RGD1563325
Chromosome Location:
Chr.7: 24647083 - 24668381 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01503181_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmem263
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001108739.1 NP_001102209.1 1 - 2 237 150
XM_008765244.1 XP_008763466.1 1 - 2 860 150

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700002343.rn.1

Additional Information:

This assay is in the 5'UTR region of the NM_001108739.1,XM_008765244.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700018720.rn.1 TC0700002343.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700018721.rn.1 TC0700002343.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0