This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01503362_m1
See other GLT8D2 GE Assays ›
Gene Symbol:
Glt8d2
Gene Name:
glycosyltransferase 8 domain containing 2
Gene Aliases:
RGD1560432
Chromosome Location:
Chr.7: 27146398 - 27196689 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01503362_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Glt8d2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008765296.1 XP_008763518.1 2 - 3 688 78
XM_008765297.1 XP_008763519.1 2 - 3 687 78
XM_008765298.1 XP_008763520.1 2 - 3 687 78
XM_008765299.1 XP_008763521.1 2 - 3 687 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008765296.1, XM_008765297.1, XM_008765298.1, XM_008765299.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0