This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01504455_m1
See other FOXRED2 GE Assays ›
Gene Symbol:
Foxred2
Gene Name:
FAD-dependent oxidoreductase domain containing 2
Gene Aliases:
RGD1306763
Chromosome Location:
Chr.7: 119172194 - 119186670 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01504455_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Foxred2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001191787.1 NP_001178716.1 7 - 8 1760 73
XM_006241932.2 XP_006241994.1 8 - 9 2806 73
XM_008765712.1 XP_008763934.1 8 - 9 2717 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700003443.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

ER-associated ubiquitin-dependent protein catabolic process
glycoprotein binding
flavin adenine dinucleotide binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019087 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019087 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019087 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019087 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P316819 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700027024.rn.1 TC0700003443.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027025.rn.1 TC0700003443.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027028.rn.1 TC0700003443.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027029.rn.1 TC0700003443.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027026.rn.1 TC0700003443.rn.1 RTA TC
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21745368 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356594 - -
RatRef-12 Expression BeadChip ILMN_59632 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0