This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01504460_g1
See other RAC2 GE Assays ›
Gene Symbol:
Rac2
Gene Name:
ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2)
Gene Aliases:
-
Chromosome Location:
Chr.7: 119783872 - 119797111 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01504460_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rac2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001008384.1 NP_001008385.1 3 - 4 320 60
XM_006242028.2 XP_006242090.1 3 - 4 363 60
GenBank mRNA BC086399.1 - 3 - 4 320 7303112 60
FQ213630.1 - 3 - 4 319 60
FQ225192.1 - 3 - 4 324 60
FQ230979.1 - 3 - 4 362 60
FQ232209.1 - 3 - 4 344 60
FQ234815.1 - 3 - 4 344 60
FQ234871.1 - 3 - 4 297 60

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rac2

Panther Classification:

Molecular Function -

small GTPase

Gene Ontology Categories:

Function(s) Process(es)

chemotaxis
actin filament organization
G-protein coupled receptor signaling pathway
small GTPase mediated signal transduction
metabolic process
positive regulation of cell proliferation
positive regulation of lamellipodium assembly
regulation of cell-substrate adhesion
cell projection assembly
actin cytoskeleton organization
bone resorption
regulation of respiratory burst
lymphocyte aggregation
positive regulation of neutrophil chemotaxis
regulation of neutrophil migration
GTPase activity
GTP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700027221.rn.1 TC0700003460.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027218.rn.1 TC0700003460.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027215.rn.1 TC0700003460.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027220.rn.1 TC0700003460.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027216.rn.1 TC0700003460.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700027223.rn.1 TC0700003460.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0