This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01504467_m1
See other CYB5R3 GE Assays ›
Gene Symbol:
Cyb5r3
Gene Name:
cytochrome b5 reductase 3
Gene Aliases:
Dia1, Nadhcb5, RNNADHCB5
Chromosome Location:
Chr.7: 124024003 - 124041564 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01504467_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cyb5r3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138877.1 NP_620232.1 7 - 8 666 67
XM_006242065.2 XP_006242127.1 7 - 8 694 67
GenBank mRNA BC062066.1 - 7 - 8 682 5599255 67
D00636.1 - 7 - 8 666 67
J03867.1 - 7 - 8 661 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700003589.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cyb5r3

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

cholesterol biosynthetic process
biological_process
oxidation-reduction process
cytochrome-b5 reductase activity, acting on NAD(P)H
AMP binding
ADP binding
flavin adenine dinucleotide binding
NAD binding
FAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700028370.rn.1 TC0700003589.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0