This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01504665_m1
See other TRPS1 GE Assays ›
Gene Symbol:
Trps1
Gene Name:
trichorhinophalangeal syndrome I
Gene Aliases:
-
Chromosome Location:
Chr.7: 90090828 - 90318748 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01504665_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134837.1 NP_001128309.1 4 - 5 2250 75
XM_008765496.1 XP_008763718.1 3 - 4 2439 75
XM_008765497.1 XP_008763719.1 3 - 4 3260 75
XM_008765498.1 XP_008763720.1 3 - 4 2104 75
XM_008765499.1 XP_008763721.1 4 - 5 1839 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700003104.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor C2H2 zinc finger transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transmembrane receptor protein serine/threonine kinase signaling pathway
biological_process
regulation of histone deacetylation
regulation of chondrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
molecular_function
transcription factor activity, sequence-specific DNA binding
zinc ion binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700023860.rn.1 TC0700003104.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700023861.rn.1 TC0700003104.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0