This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01505077_g1
See other POLR3H GE Assays ›
Gene Symbol:
Polr3h
Gene Name:
polymerase (RNA) III (DNA directed) polypeptide H
Gene Aliases:
-
Chromosome Location:
Chr.7: 123146248 - 123156558 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01505077_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr3h
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001127295.2 NP_001120767.1 1 - 2 32 120
GenBank mRNA BC167088.1 - 1 - 2 32 5621972 120
FQ220736.1 - 1 - 2 66 120
FQ225628.1 - 1 - 2 32 120
FQ229831.1 - 1 - 2 67 120

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC167088.1, NM_001127295.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
transcription from RNA polymerase III promoter
transcription initiation from RNA polymerase III promoter
biological_process
RNA polymerase III activity
molecular_function
DNA binding
DNA-directed RNA polymerase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0700033460.rn.1 TC0700004062.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700033457.rn.1 TC0700004062.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0