This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01507022_m1
See other IL4R GE Assays ›
Gene Symbol:
Il4r
Gene Name:
interleukin 4 receptor
Gene Aliases:
Il4ra
Chromosome Location:
Chr.1: 196942345 - 196967221 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01507022_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il4r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133380.2 NP_596871.2 2 - 3 156 70
XM_006230215.2 XP_006230277.1 3 - 4 307 70
XM_008759862.1 XP_008758084.1 2 - 3 5515 70
XM_008759863.1 XP_008758085.1 3 - 4 5462 70
XM_008759864.1 XP_008758086.1 3 - 4 5462 70
GenBank mRNA AB015746.1 - 2 - 3 163 70
AB015747.1 - 2 - 3 163 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AB015746.1, AB015747.1, NM_133380.2, XM_006230215.2, XM_008759862.1, XM_008759863.1, XM_008759864.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il4r

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

production of molecular mediator involved in inflammatory response
positive regulation of immunoglobulin production
inflammatory response
immune response
immunoglobulin mediated immune response
ovulation
interleukin-4-mediated signaling pathway
regulation of cell proliferation
defense response to protozoan
positive regulation of macrophage activation
positive regulation of mast cell degranulation
response to estrogen
negative regulation of T-helper 1 cell differentiation
positive regulation of T-helper 2 cell differentiation
positive regulation of chemokine secretion
positive regulation of myoblast fusion
interleukin-4 receptor activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100019885.rn.1 TC0100002575.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0