This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01507027_m1
See other IL4R GE Assays ›
Gene Symbol:
Il4r
Gene Name:
interleukin 4 receptor
Gene Aliases:
Il4ra
Chromosome Location:
Chr.1: 196942345 - 196967221 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01507027_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Il4r
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133380.2 NP_596871.2 8 - 9 938 82
XM_006230215.2 XP_006230277.1 9 - 10 1089 82
XM_006230216.2 XP_006230278.1 7 - 8 879 82
XM_008759862.1 XP_008758084.1 8 - 9 6297 82
XM_008759863.1 XP_008758085.1 9 - 10 6244 82
XM_008759864.1 XP_008758086.1 9 - 10 6244 82
GenBank mRNA AB015746.1 - 9 - 10 1020 82
AB015747.1 - 8 - 9 945 82
X69903.1 - 776 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100002575.rn.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Il4r

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

production of molecular mediator involved in inflammatory response
positive regulation of immunoglobulin production
inflammatory response
immune response
immunoglobulin mediated immune response
ovulation
interleukin-4-mediated signaling pathway
regulation of cell proliferation
defense response to protozoan
positive regulation of macrophage activation
positive regulation of mast cell degranulation
response to estrogen
negative regulation of T-helper 1 cell differentiation
positive regulation of T-helper 2 cell differentiation
positive regulation of chemokine secretion
positive regulation of myoblast fusion
interleukin-4 receptor activity
protein heterodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P089531 - -
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P337300 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 490948 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 675363 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 126444 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 915970 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 828811 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 273945 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 321424 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 911414 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 943573 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 594226 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 375660 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 680547 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 855612 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 618463 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 304298 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 442720 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 405950 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 609354 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 311018 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 48430 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 128708 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 67030 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 87053 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168415 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089531 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337300 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P337300 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P089531 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P089531 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P337300 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P337300 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700606657 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 79914 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 48746 - -
Agilent Rat cDNA Microarray (G4105A) 700606657 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10710791 - -
Affymetrix Rat Gene 1.0 ST Array 10710789 - -
Affymetrix Rat Gene 1.0 ST Array 10710787 - -
Affymetrix Rat Gene 1.0 ST Array 10710794 - -
Affymetrix Rat Gene 1.0 ST Array 10710783 - -
Affymetrix Rat Gene 1.0 ST Array 10710788 - -
Affymetrix Rat Gene 1.0 ST Array 10710790 - -
Affymetrix Rat Gene 1.0 ST Array 10710785 - -
Affymetrix Rat Gene 1.0 ST Array 10710792 - -
Affymetrix Rat Gene 1.0 ST Array 10710793 - -
Affymetrix Rat Gene 1.0 ST Array 10710786 - -
Affymetrix Rat Gene 1.0 ST Array 10710784 - -
Affymetrix Rat Gene 1.1 ST Array 10710782 - -
Rat Genome 230 2.0 Array 1368134_a_at - -
Rat Genome 230 2.0 Array 1368134_PM_a_at - -
Affymetrix HT Rat Focus Array 1368134_PM_a_at - -
Affymetrix HT Rat Focus Array 1368134_a_at - -
Affymetrix Rat Clariom S Assay PSR0100019893.rn.1 TC0100002575.rn.1 RTA TC
Rat Expression Set 230 1368134_PM_a_at - -
Rat Expression Set 230 1368134_a_at - -
Affymetrix Rat Gene 1.0 ST Array 10710782 - -
Rat Genome U34 Set X69903_at - -
Rat Genome U34 Set X69903_at - -
Rat Genome U34 Set X69903_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21649854 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1372125 - -
RatRef-12 Expression BeadChip ILMN_55821 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0