This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01508544_g1
See other NUPR1 GE Assays ›
Gene Symbol:
Nupr1 NEWGENE_69363
Gene Name:
nuclear protein, transcriptional regulator, 1
nuclear protein, transcriptional regulator, 1
Gene Aliases:
p8
Nupr1
Chromosome Location:
Chr.1: 194767484 - 194769519 on Build Rnor_6.0
Chromosome Location:
Chr.1: 198043117 - 198045203 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01508544_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nupr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053611.2 NP_446063.1 1 - 2 195 64
GenBank mRNA AA685246.1 - 1 - 2 195 64
AF014503.1 - 1 - 2 163 64
Gene Symbol: NEWGENE_69363
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008759924.1 XP_008758146.1 1 - 2 246 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006586.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nupr1
Click here to find siRNAs for Gene NEWGENE_69363

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0100051652.rn.1 TC0100006586.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100051653.rn.1 TC0100006586.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100051654.rn.1 TC0100006586.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0