This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01508954_g1
See other DCHS1 GE Assays ›
Gene Symbol:
Dchs1
Gene Name:
dachsous cadherin-related 1
Gene Aliases:
Pcdh16
Chromosome Location:
Chr.1: 170594981 - 170629062 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01508954_g1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dchs1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107544.2 NP_001101014.2 3 - 4 2309 55
XM_006229942.2 XP_006230004.1 4 - 5 2498 55
XM_008759732.1 XP_008757954.1 4 - 5 5220 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0100006301.rn.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

branching involved in ureteric bud morphogenesis
kidney development
heart morphogenesis
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
neural tube development
neurogenesis
hippo signaling
post-anal tail morphogenesis
ossification involved in bone maturation
digestive tract development
cochlea development
calcium ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P542744 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 917349 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 182743 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 910042 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 8780 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 747471 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 709672 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 285430 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 361358 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 44855 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 748287 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 861813 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 273799 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 142044 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 203158 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 509774 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 955964 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 82252 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 96284 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P542744 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P542744 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P542744 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P542744 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10724545 - -
Affymetrix Rat Gene 1.0 ST Array 10724543 - -
Affymetrix Rat Gene 1.0 ST Array 10724546 - -
Affymetrix Rat Gene 1.0 ST Array 10724548 - -
Affymetrix Rat Gene 1.0 ST Array 10724552 - -
Affymetrix Rat Gene 1.0 ST Array 10724559 - -
Affymetrix Rat Gene 1.0 ST Array 10724555 - -
Affymetrix Rat Gene 1.0 ST Array 10724550 - -
Affymetrix Rat Gene 1.0 ST Array 10724558 - -
Affymetrix Rat Gene 1.0 ST Array 10724561 - -
Affymetrix Rat Gene 1.0 ST Array 10724560 - -
Affymetrix Rat Gene 1.0 ST Array 10724557 - -
Affymetrix Rat Gene 1.0 ST Array 10724556 - -
Affymetrix Rat Gene 1.0 ST Array 10724566 - -
Affymetrix Rat Gene 1.0 ST Array 10724562 - -
Affymetrix Rat Gene 1.0 ST Array 10724563 - -
Affymetrix Rat Gene 1.0 ST Array 10724553 - -
Affymetrix Rat Gene 1.0 ST Array 10724551 - -
Affymetrix Rat Gene 1.0 ST Array 10724564 - -
Affymetrix Rat Gene 1.0 ST Array 10724544 - -
Affymetrix Rat Gene 1.0 ST Array 10724549 - -
Affymetrix Rat Gene 1.1 ST Array 10724542 - -
Rat Genome 230 2.0 Array 1386422_at - -
Rat Genome 230 2.0 Array 1388657_at - -
Rat Genome 230 2.0 Array 1386422_PM_at - -
Rat Genome 230 2.0 Array 1388657_PM_at - -
Affymetrix HT Rat Focus Array 1386422_at - -
Affymetrix HT Rat Focus Array 1388657_PM_at - -
Affymetrix HT Rat Focus Array 1388657_at - -
Affymetrix HT Rat Focus Array 1386422_PM_at - -
Affymetrix Rat Clariom S Assay PSR0100049770.rn.1 TC0100006301.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0100049771.rn.1 TC0100006301.rn.1 RTA TC
Rat Expression Set 230 1388657_at - -
Rat Expression Set 230 1388657_PM_at - -
Rat Expression Set 230 1386422_at - -
Rat Expression Set 230 1386422_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10724542 - -
Rat Genome U34 Set rc_H31750_at - -
Rat Genome U34 Set rc_AI060199_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21495468 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1351014 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0