This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01509509_g1
See other NDUFA12 GE Assays ›
Gene Symbol:
Ndufa12
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12
Gene Aliases:
RGD1311462
Chromosome Location:
Chr.7: 35125516 - 35163182 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01509509_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106781.2 NP_001100251.2 2 - 3 193 123
GenBank mRNA BF555697.1 - 2 - 3 168 123
BQ205274.1 - 123
FQ212338.1 - 2 - 3 178 123
FQ217295.1 - 2 - 3 179 123
FQ224477.1 - 2 - 3 179 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0700000526.rn.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
oxidation-reduction process
NADH dehydrogenase (ubiquinone) activity
electron carrier activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P108742 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 588627 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 430631 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 153991 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 74114 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 809680 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 184010 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 258390 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 741933 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 832165 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 136356 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 379719 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 816598 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 822799 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 414153 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 614870 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 10712 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 16593 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 169691 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108742 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P108742 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P108742 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P377784 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 80105 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10894996 - -
Affymetrix Rat Gene 1.0 ST Array 10894995 - -
Affymetrix Rat Gene 1.0 ST Array 10894998 - -
Affymetrix Rat Gene 1.0 ST Array 10882891 - -
Affymetrix Rat Gene 1.0 ST Array 10894997 - -
Affymetrix Rat Gene 1.1 ST Array 10894993 - -
Affymetrix Rat Gene 1.1 ST Array 10882890 - -
Rat Genome 230 2.0 Array 1371605_PM_at - -
Rat Genome 230 2.0 Array 1371605_at - -
Rat Genome 230 2.0 Array 1391126_at - -
Rat Genome 230 2.0 Array 1391126_PM_at - -
Affymetrix HT Rat Focus Array 1371605_PM_at - -
Affymetrix HT Rat Focus Array 1371605_at - -
Affymetrix Rat Clariom S Assay PSR0700004430.rn.1 TC0700000526.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0700004429.rn.1 TC0700000526.rn.1 RTA TC
Rat Expression Set 230 1371605_PM_at - -
Rat Expression Set 230 1371605_at - -
Rat Expression Set 230 1391126_PM_at - -
Rat Expression Set 230 1391126_at - -
Affymetrix Rat Gene 1.0 ST Array 10894993 - -
Rat Genome U34 Set rc_AI103851_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21468764 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1356229 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0