This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01509708_m1
See other MSRB3 GE Assays ›
Gene Symbol:
Msrb3
Gene Name:
methionine sulfoxide reductase B3
Gene Aliases:
-
Chromosome Location:
Chr.7: 62850506 - 62972480 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01509708_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Msrb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001055430.5 XP_001055430.4 2 - 3 732 132
XM_006241414.2 XP_006241476.1 3 - 4 326 132
XM_008765427.1 XP_008763649.1 2 - 3 752 132
XM_008765428.1 XP_008763650.1 3 - 4 376 132

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

response to oxidative stress
protein repair
oxidation-reduction process
zinc ion binding
peptide-methionine (R)-S-oxide reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0