This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01511513_g1
See other ENPP4 GE Assays ›
Gene Symbol:
Enpp4
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases:
-
Chromosome Location:
Chr.9: 19446893 - 19457946 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01511513_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Enpp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106892.1 NP_001100362.1 2 - 3 1053 71
XM_006244598.2 XP_006244660.1 3 - 4 2449 71
XM_006244600.2 XP_006244662.1 2 - 3 2334 71
XM_006244601.2 XP_006244663.1 4 - 5 2098 71
XM_006244602.2 XP_006244664.1 2 - 3 1035 71
XM_008766852.1 XP_008765074.1 3 - 4 1149 71
XM_008766853.1 XP_008765075.1 2 - 3 943 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900000276.rn.1

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
positive regulation of blood coagulation
purine ribonucleoside catabolic process
bis(5'-adenosyl)-triphosphatase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900002497.rn.1 TC0900000276.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002498.rn.1 TC0900000276.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002499.rn.1 TC0900000276.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0