This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01511514_m1
See other ENPP4 GE Assays ›
Gene Symbol:
Enpp4
Gene Name:
ectonucleotide pyrophosphatase/phosphodiesterase 4
Gene Aliases:
-
Chromosome Location:
Chr.9: 19446893 - 19457946 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01511514_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Enpp4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106892.1 NP_001100362.1 3 - 4 1225 64
XM_006244598.2 XP_006244660.1 4 - 5 2621 64
XM_006244600.2 XP_006244662.1 3 - 4 2506 64
XM_006244601.2 XP_006244663.1 5 - 6 2270 64
XM_006244602.2 XP_006244664.1 3 - 4 1207 64
XM_008766852.1 XP_008765074.1 4 - 5 1321 64
XM_008766853.1 XP_008765075.1 3 - 4 1115 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0900000276.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
positive regulation of blood coagulation
purine ribonucleoside catabolic process
bis(5'-adenosyl)-triphosphatase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P272393 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 471276 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 476797 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 669175 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 360174 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 84508 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 609323 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 61687 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P272393 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14470 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P272393 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14470 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14470 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P272393 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P272393 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 17487 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10921915 - -
Affymetrix Rat Gene 1.0 ST Array 10921914 - -
Affymetrix Rat Gene 1.0 ST Array 10921913 - -
Affymetrix Rat Gene 1.0 ST Array 10921911 - -
Affymetrix Rat Gene 1.1 ST Array 10921910 - -
Rat Genome 230 2.0 Array 1373912_PM_at - -
Rat Genome 230 2.0 Array 1373912_at - -
Affymetrix HT Rat Focus Array 1373912_at - -
Affymetrix HT Rat Focus Array 1373912_PM_at - -
Affymetrix Rat Clariom S Assay PSR0900002501.rn.1 TC0900000276.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002503.rn.1 TC0900000276.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900002502.rn.1 TC0900000276.rn.1 RTA TC
Rat Expression Set 230 1373912_PM_at - -
Rat Expression Set 230 1373912_at - -
Affymetrix Rat Gene 1.0 ST Array 10921910 - -
Rat Genome U34 Set AA851065_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22362080 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1353344 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0