This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01512736_m1
See other UBE2E3 GE Assays ›
Gene Symbol:
Ube2e3
Gene Name:
ubiquitin-conjugating enzyme E2E 3
Gene Aliases:
-
Chromosome Location:
Chr.3: 65815076 - 65871374 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01512736_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ube2e3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001047857.3 NP_001041322.2 2 - 3 568 80
XM_006234405.2 XP_006234467.1 2 - 3 495 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000921.rn.1

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
protein K63-linked ubiquitination
protein K48-linked ubiquitination
protein K11-linked ubiquitination
ubiquitin-protein transferase activity
ligase activity
ubiquitin conjugating enzyme activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0300006806.rn.1 TC0300000921.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006798.rn.1 TC0300000921.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006805.rn.1 TC0300000921.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006797.rn.1 TC0300000921.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0