This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01513845_m1
See other PPP1R1C GE Assays ›
Gene Symbol:
Ppp1r1c
Gene Name:
protein phosphatase 1, regulatory (inhibitor) subunit 1C
Gene Aliases:
RGD1562095
Chromosome Location:
Chr.3: 66673071 - 66775671 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01513845_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppp1r1c
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109200.1 NP_001102670.1 4 - 5 640 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0300000938.rn.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

phosphatase inhibitor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
positive regulation of cell growth
intracellular signal transduction
positive regulation of G1/S transition of mitotic cell cycle
protein phosphatase inhibitor activity
protein serine/threonine phosphatase inhibitor activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10303 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 808440 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 550910 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 867640 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 840099 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 183572 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 620651 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 607507 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 278023 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 504049 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 600123 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 681712 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 131049 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 279450 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 791546 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 571503 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 892039 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 859548 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 290010 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 506578 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 96970 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 536308 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 306191 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 717915 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152055 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 556796 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 647159 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163669 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 315763 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 522299 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 745097 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 221473 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 755882 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 894792 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 359125 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 612392 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 341739 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 152820 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 28465 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 65846 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 680767 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 414511 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 378197 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 4873 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 256691 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 564352 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 31330 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 486400 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 924087 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 821127 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 376200 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 309825 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 876065 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 317715 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 469271 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 202759 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 243010 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 739984 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 508139 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 814566 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 55773 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 541095 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 622980 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 756789 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 877579 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 862005 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 120329 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 158662 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 143789 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 41364 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 1763 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 112192 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 11694 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 46675 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 168948 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 136981 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 28221 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 141963 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 150150 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 167214 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10303 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10303 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10303 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P868581 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 83852 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 1073 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 25074 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10837199 - -
Affymetrix Rat Gene 1.0 ST Array 10837200 - -
Affymetrix Rat Gene 1.0 ST Array 10837197 - -
Affymetrix Rat Gene 1.0 ST Array 10837198 - -
Affymetrix Rat Gene 1.0 ST Array 10837196 - -
Affymetrix Rat Gene 1.1 ST Array 10837195 - -
Rat Genome 230 2.0 Array 1393890_PM_at - -
Rat Genome 230 2.0 Array 1393890_at - -
Affymetrix HT Rat Focus Array 1393890_PM_at - -
Affymetrix HT Rat Focus Array 1393890_at - -
Affymetrix Rat Clariom S Assay PSR0300006920.rn.1 TC0300000938.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006921.rn.1 TC0300000938.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006918.rn.1 TC0300000938.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300006919.rn.1 TC0300000938.rn.1 RTA TC
Rat Expression Set 230 1393890_PM_at - -
Rat Expression Set 230 1393890_at - -
Affymetrix Rat Gene 1.0 ST Array 10837195 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21875083 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1359717 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0