This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01515018_g1
See other ARHGAP11A GE Assays ›
Gene Symbol:
Arhgap11a
Gene Name:
Rho GTPase activating protein 11A
Gene Aliases:
RGD1309107
Chromosome Location:
Chr.3: 105281827 - 105298108 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01515018_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap11a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001168524.1 NP_001161996.1 11 - 12 1741 130

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons

Panther Classification:

Molecular Function -

G-protein modulator protein-binding activity modulator GTPase-activating protein

Gene Ontology Categories:

Function(s)

signal transduction

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P525324 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525324 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P525324 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P525324 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P525324 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10848295 - -
Affymetrix Rat Gene 1.0 ST Array 10848294 - -
Affymetrix Rat Gene 1.1 ST Array 10848293 - -
Rat Genome 230 2.0 Array 1394419_at - -
Rat Genome 230 2.0 Array 1394419_PM_at - -
Affymetrix HT Rat Focus Array 1394419_at - -
Affymetrix HT Rat Focus Array 1394419_PM_at - -
Affymetrix Rat Clariom S Assay PSR0300029613.rn.1 TC0300003909.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300029615.rn.1 TC0300003909.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300029618.rn.1 TC0300003909.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300029616.rn.1 TC0300003909.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300029614.rn.1 TC0300003909.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0300029617.rn.1 TC0300003909.rn.1 RTA TC
Rat Expression Set 230 1394419_PM_at - -
Rat Expression Set 230 1394419_at - -
Affymetrix Rat Gene 1.0 ST Array 10848293 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21019642 - -
ABI Rat Genome Survey Microarray 21407450 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1353170 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0