This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01518023_m1
See other BATF GE Assays ›
Gene Symbol:
Batf
Gene Name:
basic leucine zipper transcription factor, ATF-like
Gene Aliases:
B-ATF
Chromosome Location:
Chr.6: 109562407 - 109584541 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01518023_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Batf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106748.1 NP_001100218.1 1 - 2 303 80
XM_008764758.1 XP_008762980.1 2 - 3 405 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0600001198.rn.1

Set Membership:

Amplicon length between 71 and 85 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

cytokine production
lymphoid progenitor cell differentiation
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cellular response to DNA damage stimulus
biological_process
DNA damage response, signal transduction by p53 class mediator
defense response to protozoan
myeloid dendritic cell differentiation
T-helper 2 cell differentiation
isotype switching
positive regulation of transcription from RNA polymerase II promoter
hematopoietic stem cell differentiation
T-helper 17 cell differentiation
T-helper 17 cell lineage commitment
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
molecular_function
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0600008481.rn.1 TC0600001198.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0600008482.rn.1 TC0600001198.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0