This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01518551_m1
See other EXTL2 GE Assays ›
Gene Symbol:
Extl2
Gene Name:
exostosin-like glycosyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 218951124 - 218972576 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01518551_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100704.1 NP_001094174.1 3 - 4 610 89
XM_006233215.2 XP_006233277.1 3 - 4 1155 89
XM_006233216.2 XP_006233278.1 3 - 4 896 89
XM_006233217.2 XP_006233279.1 2 - 3 687 89
GenBank mRNA BC162014.1 - 3 - 4 610 9091843 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002042.rn.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
N-acetylglucosamine metabolic process
heparan sulfate proteoglycan biosynthetic process
UDP-N-acetylgalactosamine metabolic process
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
alpha-1,4-N-acetylgalactosaminyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200014143.rn.1 TC0200002042.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rz6mw:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0