This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01518552_m1
See other EXTL2 GE Assays ›
Gene Symbol:
Extl2
Gene Name:
exostosin-like glycosyltransferase 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 218951124 - 218972576 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01518552_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100704.1 NP_001094174.1 4 - 5 684 96
XM_006233215.2 XP_006233277.1 4 - 5 1229 96
XM_006233216.2 XP_006233278.1 4 - 5 970 96
XM_006233217.2 XP_006233279.1 3 - 4 761 96
GenBank mRNA BC162014.1 - 4 - 5 684 9091843 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200002042.rn.1

Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

glycosaminoglycan biosynthetic process
N-acetylglucosamine metabolic process
heparan sulfate proteoglycan biosynthetic process
UDP-N-acetylgalactosamine metabolic process
glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
alpha-1,4-N-acetylgalactosaminyltransferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P607298 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 443072 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 855362 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 772153 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 216274 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 803355 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 830833 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 516453 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 544264 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 619039 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 143082 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 437673 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 592150 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 693396 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 260797 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 391769 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 512153 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 62590 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 110336 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P607298 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P607298 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P607298 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P607298 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P534374 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10818592 - -
Affymetrix Rat Gene 1.0 ST Array 10818593 - -
Affymetrix Rat Gene 1.0 ST Array 10818591 - -
Affymetrix Rat Gene 1.1 ST Array 10818590 - -
Rat Genome 230 2.0 Array 1379354_PM_at - -
Rat Genome 230 2.0 Array 1379354_at - -
Affymetrix HT Rat Focus Array 1379354_at - -
Affymetrix HT Rat Focus Array 1379354_PM_at - -
Affymetrix Rat Clariom S Assay PSR0200014145.rn.1 TC0200002042.rn.1 RTA TC
Rat Expression Set 230 1379354_at - -
Rat Expression Set 230 1379354_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10818590 - -
Rat Genome U34 Set rc_AA852056_at - -
Rat Genome U34 Set rc_AA851239_at - -
Rat Genome U34 Set rc_AA852056_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21339113 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1357905 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0