This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01518601_m1
See other AIMP1 GE Assays ›
Gene Symbol:
Aimp1
Gene Name:
aminoacyl tRNA synthetase complex-interacting multifunctional protein 1
Gene Aliases:
Emap2, Scye1
Chromosome Location:
Chr.2: 237727782 - 237751327 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01518601_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aimp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053757.1 NP_446209.1 4 - 5 440 102
GenBank mRNA BC098675.1 - 4 - 5 440 7322792 102
FQ226969.1 - 2 - 3 216 102
FQ229959.1 - 2 - 3 216 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200004718.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Inflammation 1700 Inventoried Rat_Inflammation Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aimp1

Panther Classification:

Molecular Function -

translational protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
inflammatory response
cell-cell signaling
negative regulation of angiogenesis
blood vessel morphogenesis
leukocyte migration
defense response to virus
tRNA binding
cytokine activity
protein homodimerization activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P136775 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 373167 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 464620 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 360735 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 529346 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 951674 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 450029 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 901607 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 445685 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 380782 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 439001 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 354199 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 658589 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 589959 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 896332 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 188310 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 78036 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 928344 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 315800 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 973036 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 381057 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 111175 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 150628 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 25588 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 71685 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136775 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P136775 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P136775 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P16995 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P735353 - -
Matsumoto (Agilent Rat cDNA Microarray G4105A) 700597475 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 8604 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 1722 - -
Agilent Rat cDNA Microarray (G4105A) 700597475 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10826864 - -
Affymetrix Rat Gene 1.0 ST Array 10826861 - -
Affymetrix Rat Gene 1.0 ST Array 10826863 - -
Affymetrix Rat Gene 1.0 ST Array 10826860 - -
Affymetrix Rat Gene 1.0 ST Array 10826865 - -
Affymetrix Rat Gene 1.0 ST Array 10826862 - -
Affymetrix Rat Gene 1.1 ST Array 10826859 - -
Rat Genome 230 2.0 Array 1374279_at - -
Rat Genome 230 2.0 Array 1374279_PM_at - -
Rat Genome 230 2.0 Array 1379810_PM_at - -
Rat Genome 230 2.0 Array 1379810_at - -
Affymetrix HT Rat Focus Array 1374279_PM_at - -
Affymetrix HT Rat Focus Array 1379810_PM_at - -
Affymetrix HT Rat Focus Array 1374279_at - -
Affymetrix HT Rat Focus Array 1379810_at - -
Affymetrix Rat Clariom S Assay PSR0200032792.rn.1 TC0200004718.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200032793.rn.1 TC0200004718.rn.1 RTA TC
Rat Expression Set 230 1374279_PM_at - -
Rat Expression Set 230 1374279_at - -
Rat Expression Set 230 1379810_PM_at - -
Rat Expression Set 230 1379810_at - -
Affymetrix Rat Gene 1.0 ST Array 10826859 - -
Rat Genome U34 Set rc_AI136838_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20753995 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1357417 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0