This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01519506_m1
See other ALDH4A1 GE Assays ›
Gene Symbol:
Aldh4a1
Gene Name:
aldehyde dehydrogenase 4 family, member A1
Gene Aliases:
-
Chromosome Location:
Chr.5: 158090238 - 158115725 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01519506_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh4a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001134698.1 NP_001128170.1 14 - 15 1610 82
GenBank mRNA BC168153.1 - 14 - 15 1610 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500001778.rn.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

glutamate biosynthetic process
proline biosynthetic process
proline catabolic process to glutamate
oxidation-reduction process
1-pyrroline-5-carboxylate dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
structural molecule activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
identical protein binding

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P164295 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 163436 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 257229 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 905492 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 140598 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 397484 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 720997 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 107415 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 584576 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 204257 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 748657 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 315563 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 731483 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 512471 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 145813 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 58649 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164296 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031640 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P031640 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P164296 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P164296 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P031640 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10873545 - -
Affymetrix Rat Gene 1.0 ST Array 10873551 - -
Affymetrix Rat Gene 1.0 ST Array 10873546 - -
Affymetrix Rat Gene 1.0 ST Array 10873550 - -
Affymetrix Rat Gene 1.0 ST Array 10873555 - -
Affymetrix Rat Gene 1.0 ST Array 10873549 - -
Affymetrix Rat Gene 1.0 ST Array 10873544 - -
Affymetrix Rat Gene 1.0 ST Array 10873556 - -
Affymetrix Rat Gene 1.0 ST Array 10873554 - -
Affymetrix Rat Gene 1.0 ST Array 10873557 - -
Affymetrix Rat Gene 1.0 ST Array 10873553 - -
Affymetrix Rat Gene 1.0 ST Array 10873547 - -
Affymetrix Rat Gene 1.0 ST Array 10873552 - -
Affymetrix Rat Gene 1.1 ST Array 10873534 - -
Rat Genome 230 2.0 Array 1372277_PM_at - -
Rat Genome 230 2.0 Array 1372277_at - -
Affymetrix HT Rat Focus Array 1372277_at - -
Affymetrix HT Rat Focus Array 1372277_PM_at - -
Affymetrix Rat Clariom S Assay PSR0500013419.rn.1 TC0500001778.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500013418.rn.1 TC0500001778.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500013420.rn.1 TC0500001778.rn.1 RTA TC
Rat Expression Set 230 1372277_at - -
Rat Expression Set 230 1372277_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10873534 - -
Rat Genome U34 Set rc_AI011510_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 20994702 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0