This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01520291_m1
See other EXTL1 GE Assays ›
Gene Symbol:
Extl1
Gene Name:
exostosin-like glycosyltransferase 1
Gene Aliases:
-
Chromosome Location:
Chr.5: 152573755 - 152588956 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01520291_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Extl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001107985.1 NP_001101455.1 9 - 10 1726 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500003813.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s)

glycosaminoglycan biosynthetic process
protein glycosylation
heparan sulfate proteoglycan biosynthetic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500028599.rn.1 TC0500003813.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0