This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01520477_m1
See other GJB3 GE Assays ›
Gene Symbol:
Gjb3
Gene Name:
gap junction protein, beta 3
Gene Aliases:
-
Chromosome Location:
Chr.5: 145390590 - 145397279 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01520477_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gjb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006238845.2 XP_006238907.1 1 - 2 1281 112

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_006238845.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

gap junction

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
placenta development
cell communication
sensory perception of sound
skin development
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline