This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01520519_g1
See other PLA2G2D GE Assays ›
Gene Symbol:
Pla2g2d
Gene Name:
phospholipase A2, group IID
Gene Aliases:
-
Chromosome Location:
Chr.5: 157222636 - 157228795 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01520519_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pla2g2d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013428.1 NP_001013446.1 2 - 3 206 67
GenBank mRNA BC079065.1 - 2 - 3 219 67
BC091221.1 - 2 - 3 206 7125035 67
FQ227390.1 - 2 - 3 183 67
FQ231588.1 - 2 - 3 183 67
FQ234103.1 - 2 - 3 183 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0500001758.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pla2g2d

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

phospholipid metabolic process
lipid catabolic process
phospholipase A2 activity
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500013073.rn.1 TC0500001758.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500013072.rn.1 TC0500001758.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500013074.rn.1 TC0500001758.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0