This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01520710_m1
See other ZCCHC17 GE Assays ›
Gene Symbol:
Zcchc17
Gene Name:
zinc finger, CCHC domain containing 17
Gene Aliases:
RGD1565267
Chromosome Location:
Chr.5: 148535519 - 148577312 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01520710_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zcchc17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109267.1 NP_001102737.1 1 - 2 84 67
XM_006238968.2 XP_006239030.1 1 - 2 104 67
XM_006238969.2 XP_006239031.1 1 - 2 98 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109267.1, XM_006238968.2, XM_006238969.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

Gene Ontology Categories:

Process(es)

nucleic acid binding
zinc ion binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0500027684.rn.1 TC0500003705.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0500027685.rn.1 TC0500003705.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0