This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01521362_g1
See other TWF2 GE Assays ›
Gene Symbol:
Twf2
Gene Name:
twinfilin actin-binding protein 2
Gene Aliases:
-
Chromosome Location:
Chr.8: 114897011 - 114909531 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01521362_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Twf2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135238.1 NP_001128710.1 1 - 2 177 117
GenBank mRNA BC158614.1 - 1 - 2 125 7386008 117

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC158614.1,NM_001135238.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of lamellipodium assembly
positive regulation of neuron projection development
phosphorylation
negative regulation of actin filament polymerization
regulation of actin cytoskeleton organization
sequestering of actin monomers
positive regulation of axon extension
barbed-end actin filament capping
cellular response to retinoic acid
cellular response to growth factor stimulus
actin monomer binding
protein kinase C binding
ATP binding
phosphatidylinositol-4,5-bisphosphate binding
kinase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0800010745.rn.1 TC0800001470.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800010744.rn.1 TC0800001470.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0800010746.rn.1 TC0800001470.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0