This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01521564_g1
See other DRAM2 GE Assays ›
Gene Symbol:
Dram2
Gene Name:
DNA-damage regulated autophagy modulator 2
Gene Aliases:
Tmem77
Chromosome Location:
Chr.2: 209160904 - 209189641 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01521564_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dram2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001025018.1 NP_001020189.1 5 - 6 550 80
XM_006233120.2 XP_006233182.1 5 - 6 494 80
XM_006233121.2 XP_006233183.1 4 - 5 434 80
XM_006233122.2 XP_006233184.1 4 - 5 535 80
XM_006233123.2 XP_006233185.1 5 - 6 496 80
GenBank mRNA BC091251.1 - 5 - 6 550 7372838 80
FQ218349.1 - 5 - 6 559 80
FQ228782.1 - 5 - 6 647 80

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dram2

Gene Ontology Categories:

Function(s)

regulation of autophagy
apoptotic nuclear changes

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200013415.rn.1 TC0200001946.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200013414.rn.1 TC0200001946.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7xkh5:80/100.66.78.74:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0