This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01521954_g1
See other ATP5F1 GE Assays ›
Gene Symbol:
Atp5f1 LOC100911417
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex, subunit B1
ATP synthase subunit b, mitochondrial-like
Gene Aliases:
-
-
Chromosome Location:
Chr.2: 208566385 - 208577147 on Build Rnor_6.0
Chromosome Location:
Chr.2: 208408890 - 208420159 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01521954_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5f1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134365.2 NP_599192.1 3 - 4 360 88
GenBank mRNA BC063808.1 - 3 - 4 301 6920021 88
FQ212544.1 - 3 - 4 296 88
FQ214336.1 - 3 - 4 288 88
FQ214353.1 - 3 - 4 322 88
FQ214371.1 - 3 - 4 296 88
FQ214449.1 - 3 - 4 259 88
FQ214672.1 - 3 - 4 295 88
FQ214784.1 - 3 - 4 294 88
FQ215244.1 - 3 - 4 259 88
FQ215480.1 - 3 - 4 259 88
FQ215823.1 - 3 - 4 312 88
FQ216062.1 - 3 - 4 261 88
FQ216247.1 - 3 - 4 88
FQ216597.1 - 3 - 4 251 88
FQ216773.1 - 3 - 4 259 88
FQ216829.1 - 3 - 4 259 88
FQ216922.1 - 3 - 4 259 88
FQ219899.1 - 3 - 4 288 88
FQ223652.1 - 3 - 4 259 88
FQ223683.1 - 3 - 4 285 88
FQ229976.1 - 3 - 4 296 88
FQ232356.1 - 3 - 4 296 88
M35052.1 - 3 - 4 279 88
Gene Symbol: LOC100911417
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003749372.2 XP_003749420.1 355 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5f1
Click here to find siRNAs for Gene LOC100911417

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP synthesis coupled proton transport
ATP metabolic process
substantia nigra development
proton-transporting ATP synthase activity, rotational mechanism
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200030429.rn.1 TC0200004408.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0