This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01523121_m1
See other SEC24B GE Assays ›
Gene Symbol:
Sec24b
Gene Name:
SEC24 family member B
Gene Aliases:
-
Chromosome Location:
Chr.2: 230286891 - 230356880 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01523121_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sec24b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001106474.1 NP_001099944.1 8 - 9 1855 58
XM_006233275.2 XP_006233337.1 8 - 9 1931 58
XM_006233276.2 XP_006233338.1 7 - 8 1825 58
XM_006233277.2 XP_006233339.1 6 - 7 1651 58
XM_008761458.1 XP_008759680.1 8 - 9 1928 58

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC0200004638.rn.1

Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s) Process(es)

neural tube closure
auditory receptor cell morphogenesis
outflow tract morphogenesis
intracellular protein transport
ER to Golgi vesicle-mediated transport
cochlear nucleus development
aorta morphogenesis
auditory receptor cell stereocilium organization
lung morphogenesis
lung lobe morphogenesis
coronary artery morphogenesis
pulmonary artery morphogenesis
cardiovascular system development
regulation of establishment of planar polarity involved in neural tube closure
regulation of cargo loading into COPII-coated vesicle
zinc ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200032247.rn.1 TC0200004638.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0