This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01523548_gH
See other GSTM1 GE Assays ›
Gene Symbol:
Gstm1
Gene Name:
glutathione S-transferase mu 1
Gene Aliases:
GSTA3
Chromosome Location:
Chr.2: 210803869 - 210809405 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01523548_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gstm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017014.1 NP_058710.1 5 - 6 399 72
GenBank mRNA BC063172.1 - 5 - 6 439 6920688 72
FJ179401.1 - 5 - 6 363 72
FQ211067.1 - 5 - 6 398 72
FQ212742.1 - 5 - 6 392 72
FQ213204.1 - 5 - 6 392 72
FQ213696.1 - 5 - 6 586 72
FQ214055.1 - 5 - 6 498 72
FQ217092.1 - 5 - 6 465 72
FQ218789.1 - 5 - 6 398 72
FQ218795.1 - 5 - 6 435 72
FQ229836.1 - 5 - 6 446 72
FQ229941.1 - 5 - 6 426 72
J02810.1 - 5 - 6 399 72
M11719.1 - 5 - 6 400 72
M28241.1 - 5 - 6 459 72
X04229.1 - 5 - 6 399 72

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gstm1

Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
sensory perception of smell
metabolic process
response to metal ion
response to lithium ion
response to lead ion
nitrobenzene metabolic process
cellular response to drug
xenobiotic catabolic process
response to drug
response to amino acid
response to ethanol
response to antibiotic
response to axon injury
cellular detoxification of nitrogen compound
glutathione transferase activity
steroid binding
nickel cation binding
enzyme binding
protein homodimerization activity
glutathione binding
protein heterodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0