This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01524470_m1
See other FAM173B GE Assays ›
Gene Symbol:
Fam173b
Gene Name:
family with sequence similarity 173, member B
Gene Aliases:
RGD1560629
Chromosome Location:
Chr.2: 84678888 - 84694152 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01524470_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam173b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109178.1 NP_001102648.1 1 - 2 228 60
XM_008760792.1 XP_008759014.1 1 - 2 288 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001109178.1, XM_008760792.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0200004451.rn.1 TC0200000722.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004450.rn.1 TC0200000722.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004443.rn.1 TC0200000722.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004444.rn.1 TC0200000722.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline