This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01524473_m1
See other FAM173B GE Assays ›
Gene Symbol:
Fam173b
Gene Name:
family with sequence similarity 173, member B
Gene Aliases:
RGD1560629
Chromosome Location:
Chr.2: 84678888 - 84694152 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01524473_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam173b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001109178.1 NP_001102648.1 4 - 5 693 88

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 3' Most Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P102876 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 286009 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 284298 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 159875 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 447360 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 142032 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 280621 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 3527 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 826366 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 287568 - -
Agilent SurePrint G3 Rat CGH Microarray 1x1M 36399 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 35438 - -
Agilent SurePrint G3 Rat CGH Microarray 4x180K 58278 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102876 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P102876 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P102876 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P102876 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 45944 - -
Agilent Rat Genome CGH Microarray 105A (G4436A) 3617 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10814115 - -
Affymetrix Rat Gene 1.0 ST Array 10814113 - -
Affymetrix Rat Gene 1.0 ST Array 10814114 - -
Affymetrix Rat Gene 1.0 ST Array 10814116 - -
Affymetrix Rat Gene 1.1 ST Array 10814111 - -
Rat Genome 230 2.0 Array 1373906_PM_at - -
Rat Genome 230 2.0 Array 1373906_at - -
Affymetrix HT Rat Focus Array 1373906_at - -
Affymetrix HT Rat Focus Array 1373906_PM_at - -
Affymetrix Rat Clariom S Assay PSR0200004456.rn.1 TC0200000722.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004457.rn.1 TC0200000722.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0200004455.rn.1 TC0200000722.rn.1 RTA TC
Rat Expression Set 230 1373906_at - -
Rat Expression Set 230 1373906_PM_at - -
Affymetrix Rat Gene 1.0 ST Array 10814111 - -
Rat Genome U34 Set rc_AI104594_at - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 22140103 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1370742 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline