This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01525970_g1
See other GPSM3 GE Assays ›
Gene Symbol:
Gpsm3
Gene Name:
G-protein signaling modulator 3
Gene Aliases:
AGS4, G18, Ng1
Chromosome Location:
Chr.20: 4355246 - 4357107 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01525970_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpsm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001003974.2 NP_001003974.1 2 - 3 445 61
GenBank mRNA BC088220.1 - 2 - 3 445 7384543 61
FQ229559.1 - 2 - 3 265 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC2000000146.rn.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpsm3

Gene Ontology Categories:

Function(s) Process(es)

immune response
regulation of G-protein coupled receptor protein signaling pathway
regulation of catalytic activity
GDP-dissociation inhibitor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR2000001322.rn.1 TC2000000146.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0