This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01527024_g1
See other ATP5A1 GE Assays ›
Gene Symbol:
Atp5a1
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
Gene Aliases:
-
Chromosome Location:
Chr.18: 74156553 - 74164490 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01527024_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp5a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023093.1 NP_075581.1 10 - 11 1471 135
GenBank mRNA BC061830.1 - 10 - 11 1471 5599635 135
FQ214031.1 - 7 - 8 883 135
FQ225416.1 - 5 - 6 786 135
FQ227531.1 - 5 - 6 786 135
FQ227720.1 - 4 - 5 606 135
J05266.1 - 10 - 11 1392 135
X56133.1 - 3 - 4 468 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1800000866.rn.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp5a1

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of endothelial cell proliferation
lipid metabolic process
ATP synthesis coupled proton transport
ATP hydrolysis coupled proton transport
ATP metabolic process
hydrogen ion transmembrane transport
ATP binding
ATPase activity
MHC class I protein binding
ADP binding
poly(A) RNA binding
proton-transporting ATP synthase activity, rotational mechanism
proton-transporting ATPase activity, rotational mechanism

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR1800006265.rn.1 TC1800000866.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0