This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01527780_g1
See other NDUFA11 GE Assays ›
Gene Symbol:
Ndufa11
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 11
Gene Aliases:
-
Chromosome Location:
Chr.9: 10334104 - 10338163 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01527780_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_212517.1 NP_997682.1 2 - 3 239 58
GenBank mRNA AY272059.1 - 2 - 3 239 58
FQ209810.1 - 2 - 3 219 58

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufa11

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s)

oxidation-reduction process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Rat Clariom S Assay PSR0900012061.rn.1 TC0900001462.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900012062.rn.1 TC0900001462.rn.1 RTA TC
Affymetrix Rat Clariom S Assay PSR0900012060.rn.1 TC0900001462.rn.1 RTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-l2zpb:80/100.66.75.138:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0