This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01528116_m1
See other NXPE4 GE Assays ›
Gene Symbol:
Nxpe4 Nxpe5
Gene Name:
neurexophilin and PC-esterase domain family, member 4
neurexophilin and PC-esterase domain family, member 5
Gene Aliases:
-
RGD1559954
Chromosome Location:
Chr.12: 19428950 - 19451575 on Build Rnor_6.0
Chromosome Location:
Chr.12: 19518859 - 19530862 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
-
Assay ID Rn01528116_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nxpe4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001056892.5 XP_001056892.3 4 - 5 1343 123
XM_006249027.2 XP_006249089.1 5 - 6 1197 123
XM_006249028.2 XP_006249090.1 4 - 5 1429 123
Gene Symbol: Nxpe5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008769074.1 XP_008767296.1 3 - 4 1385 123

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Rattus norvegicus 1700 Probe spans exons

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P540609 - -
Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P540609 - -
Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P540609 - -
SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P540609 - -
Array Name Probe IDs Targets Target Type
Affymetrix Rat Gene 1.0 ST Array 10757252 - -
Affymetrix Rat Gene 1.1 ST Array 10757249 - -
Affymetrix Rat Gene 1.0 ST Array 10757249 - -
Array Name Probe IDs Targets Target Type
ABI Rat Genome Survey Microarray 21923838 - -
Array Name Probe IDs Targets Target Type
Illumina ratRef-12 v1.0 expression beadchip ILMN_1361917 - -
Illumina ratRef-12 v1.0 expression beadchip ILMN_1651161 - -
RatRef-12 Expression BeadChip ILMN_58416 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0