This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rn01528302_g1
See other FTSJ1 GE Assays ›
Gene Symbol:
Ftsj1
Gene Name:
FtsJ RNA methyltransferase homolog 1 (E. coli)
Gene Aliases:
RGD1561061
Chromosome Location:
Chr.X: 14993714 - 15005996 on Build Rnor_6.0
Species:
Rat
Species Specific ID (Flybase ID):
-
Assay ID Rn01528302_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ftsj1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006256749.2 XP_006256811.1 8 - 9 1026 108
XM_006256750.2 XP_006256812.1 8 - 9 1026 108
XM_006256753.2 XP_006256815.1 8 - 9 874 108
XM_008773086.1 XP_008771308.1 8 - 9 1247 108
XM_008773087.1 XP_008771309.1 8 - 9 1023 108
XM_008773088.1 XP_008771310.1 8 - 9 650 108
XM_008773090.1 XP_008771312.1 8 - 9 873 108
XM_008773091.1 XP_008771313.1 8 - 9 1032 108
XM_008773092.1 XP_008771314.1 4 - 5 1940 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA nucleoside ribose methylation
cytoplasmic translation
tRNA methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zl4nj:80/100.66.77.81:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0